Learn More
M-CSF Monoclonal Antibody (26730), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523815
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse M-CSF. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
M-CSF Monoclonal antibody specifically detects M-CSF in Human samples. It is validated for Immunocytochemistry, Neutralization, Western Blot.Specifications
M-CSF | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P09603 | |
Csf1 | |
E. coli-derived recombinant human M-CSF | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Neutralization, Western Blot | |
26730 | |
Unconjugated | |
Csf1 | |
C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1 | |
Mouse | |
Protein A/G | |
RUO | |
1435 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |