Learn More
MCP-1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547200
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) CCL2/MCP-1 is observed, approximately 10% cross-reactivity with recombinant mouse (rm) JE, recombinant rat JE, and recombinant cotton rat JE is observed, and less than 5% cross-reactivity with rhEotaxin, rmMARC, and rhMCP-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
MCP-1 Polyclonal antibody specifically detects MCP-1 in Canine samples. It is validated for Immunocytochemistry, Neutralization, Western Blot.Specifications
MCP-1 | |
Polyclonal | |
Unconjugated | |
Ccl2 | |
Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF | |
Goat | |
Antigen affinity chromatography | |
RUO | |
403981 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P52203 | |
Ccl2 | |
E. coli-derived recombinant canine CCL2/JE/MCP-1 Gln24-Pro101 | |
100 μg | |
Primary | |
Canine | |
Antibody | |
IgG |