Learn More
MEK2 Recombinant Rabbit Monoclonal Antibody (B19H36L8), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 700829
Additional Details : Weight : 1.00000kg
Description
This antibody is predicted to react with bovine, chicken, mouse and rat based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
MEK2 Recombinant Monoclonal antibody specifically detects MEK2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
MEK2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
AA589381; CFC4; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 2; ERK kinase 2; FLJ26075; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP2K1; MAP2K2; MAPK; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MAPKK2; MEK 1; MEK 2; MEK-1; Mek2; MEK-2; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; MK2; MKK1; Mkk2; OTTHUMP00000165827; PRKMK1; PRKMK2; protein kinase, mitogen activated, kinase 2, p45 | |
proprietary | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Western Blot | |
B19H36L8 | |
Unconjugated | |
P36507 | |
Rabbit | |
Protein A | |
RUO | |
5605 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |