missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129852
Additional Details : Weight : 0.01000kg
Description
PA1-29852 detects MEK2 from human samples.
MEK2 Polyclonal antibody specifically detects MEK2 in Human samples. It is validated for Western Blot.Specifications
MEK2 | |
Polyclonal | |
Unconjugated | |
MAP2K2 | |
AA589381; CFC4; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 2; ERK kinase 2; FLJ26075; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP2K1; MAP2K2; MAPK; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MAPKK2; MEK 1; MEK 2; MEK-1; Mek2; MEK-2; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; MK2; MKK1; Mkk2; OTTHUMP00000165827; PRKMK1; PRKMK2; protein kinase, mitogen activated, kinase 2, p45 | |
Rabbit | |
IgG fraction | |
RUO | |
5605 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
P36507 | |
MAP2K2 | |
Synthetic peptide (Human). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |