Learn More
DIS3L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541948
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: WKVVKPESND KETEAAYESD IPEELCGHHL PQQSLKSYND SPDVIVEAQF Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 79%; Rat: 86%
DIS3L2 Polyclonal antibody specifically detects DIS3L2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
DIS3L2 | |
Polyclonal | |
Unconjugated | |
DIS3L2 | |
4930429A22Rik; 8030493P09Rik; DIS3 like 3'-5' exoribonuclease 2; DIS3 mitotic control homolog (S. cerevisiae)-like 2; DIS3 mitotic control homolog-like 2; DIS3 mitotic control-like 2; Dis3l2; DIS3-like 3'-5' exoribonuclease 2; DIS3-like exonuclease 2; FAM6A; family with sequence similarity 6, member A; hDIS3L2; PRLMNS; RGD1560168 | |
Rabbit | |
Protein A | |
RUO | |
129563 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IYB7 | |
DIS3L2 | |
Synthetic peptide directed towards the N-terminal of human MGC42174 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |