Learn More
HLA Class I Monoclonal Antibody (2G5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180907
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Mouse MHC Class I antibody, clone 2G5 recognizes a monomorphic epitope present on murine MHC class I molecules, expressed at varying levels on the majority of nucleated cells.
HLA Class I Monoclonal antibody specifically detects HLA Class I in Bovine, Guinea Pig, Hamster, Human, Mouse, Ovine, Porcine, Rat samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
HLA Class I | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01889, P04439, P10321 | |
HLA-A, HLA-B, HLA-C, LOC100515902 | |
Purified H-2Kb and H-2Db MHC-I molecules. | |
250 μg | |
Primary | |
Bovine, Guinea Pig, Hamster, Human, Mouse, Ovine, Porcine, Rat | |
Antibody | |
IgG2b |
Flow Cytometry, Immunoprecipitation | |
2G5 | |
Unconjugated | |
HLA-C | |
A29 protein; alpha chain of MHC class II; antigen presenting molecule; AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; class I MHC, HLA-B antigen; D6S204; epididymis secretory protein Li 83; H-2K; H2-K; H-2K(d); HEL-S-83; histocompatibility antigen; HLA A; HLA -B; HLA B-40011; HLA B44; HLA C; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen C alpha chain; HLA class I histocompatibility antigen Cw-1 alpha chain; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA class I histocompatibility a | |
Mouse | |
Protein A | |
RUO | |
100515902, 3105, 3106, 3107 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |