Learn More
HLA-ABC Monoclonal Antibody (W6/32), Functional Grade, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180123
7125.00 SEK valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
This antibody does not react with amphibia, chicken, dog, goat, guinea pig, mouse or rabbit. The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control for immunohistochemical applications is tonsil. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. This product is a low-endotoxin, preservative-free formulation. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Mouse anti Human HLA ABC antibody, clone W6/32 recognizes an antigenic determinant shared among products of the HLA A, B and C loci. Clone W6/32 recognizes a conformational epitope, reacting with HLA class I alpha3 and alpha2 domains.
HLA-ABC Monoclonal antibody specifically detects HLA-ABC in Bovine, Human, Non-human Primate, Rodent samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
HLA-ABC | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P01889, P04439, P10321 | |
HLA-A, HLA-B, HLA-C | |
Purified human tonsil lymphocyte membranes. | |
500 μL | |
Primary | |
Bovine, Human, Non-human Primate, Rodent | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation | |
W6/32 | |
Functional Grade | |
HLA-C | |
A29 protein; alpha chain of MHC class II; antigen presenting molecule; AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; class I MHC, HLA-B antigen; D6S204; epididymis secretory protein Li 83; H-2K; H2-K; H-2K(d); HEL-S-83; histocompatibility antigen; HLA A; HLA -B; HLA B-40011; HLA B44; HLA C; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen C alpha chain; HLA class I histocompatibility antigen Cw-1 alpha chain; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-11 alpha chain-like; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibil | |
Mouse | |
Protein G | |
RUO | |
3105, 3106, 3107 | |
4° C, do not freeze | |
Liquid |