Learn More
HLA-E Monoclonal Antibody (MEM-E/08), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119362
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. It shows only with weak cross-reactivity with following classical MHC Class I molecules (MHC Class Ia): HLA-A24, -B7, -B27, -B51, -B54, -C7.
HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
HLA-E | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P13747 | |
HLA-E | |
Recombinant HLA-E refolded with beta-2 Microglobulin and peptide. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
MEM-E/08 | |
Unconjugated | |
HLA-E | |
A29 protein; antigen presenting molecule; Aw-33; Aw-74; DAMA-277I14.1; DAQB-90C11.16; DKFZp686P19218; EA1.2; EA2.1; FLJ26655; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, alpha chain E; HLA class I histocompatibility antigen, E alpha chain; HLA locus; HLA-6.2; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; HLAE; HLA-E; Human leukocyte antigen A; leukocyte antigen class I-A; lymphocyte antigen; major histocompatibility complex, class I, A; major histocompatibility complex, class I, E; MCH class I antigen; MHC; MHC 1; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; M | |
Mouse | |
Protein A | |
RUO | |
3133 | |
4° C, do not freeze | |
Liquid |