Learn More
HLA-DR/DP/DQ Monoclonal Antibody (WR18), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180678
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for immunohistochemical applications is tonsil. Removal of sodium azide by dialysis is recommended prior to use in functional assays. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human HLA DP DQ DR antibody, clone WR18 reacts with a monomorphic determinant common to DP, DQ and DR beta chains, which are expressed by antigen presenting cells, B cells, monocytes and activated T lymphocytes.
HLA-DR/DP/DQ Monoclonal antibody specifically detects HLA-DR/DP/DQ in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin).Specifications
HLA-DR/DP/DQ | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P01911, P01920, P04440 | |
HLA-DPB1, HLA-DQB1, HLA-DRB1 | |
Human HLA Class II (DP, DQ, DR). | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin) | |
WR18 | |
Unconjugated | |
HLA-DQB1 | |
beta1 domain MHC class II HLA DPB; CELIAC1; class II HLA beta chain; Clone P2-beta-3; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DR beta; DLA-DRB1; DLA-DRBB1; DPB1; DR-16; DRB; DRB1; DRBB1; DRw10; DW2.2/DR2.2; FLJ75017; FLJ76359; HLA class II antigen; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA DP14-beta chain; HLA-D; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DQ; HLA-DQB; HLA-DQB1; HLA-DR; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; IDDM1; integral membrane glycoprotein; LA-DRB; lymphocyte antigen; lymphocyte a | |
Mouse | |
Protein G | |
RUO | |
3115, 3119, 3123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |