missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR/DP Monoclonal Antibody (HL-38), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119145
Additional Details : Weight : 0.01000kg
Description
Western Blot: non-reducing conditions.
HLA-DR/DP Monoclonal antibody specifically detects HLA-DR/DP in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
HLA-DR/DP | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P01911, P04440 | |
HLA-DPB1, HLA-DRB1 | |
Burkitt's lymphoma cell line Raji. | |
100 μg | |
Primary | |
Human, Porcine | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
HL-38 | |
Unconjugated | |
HLA-DPB1 | |
beta1 domain MHC class II HLA DPB; class II HLA beta chain; Clone P2-beta-3; DPB1; DR-16; DRB1; DRw10; DW2.2/DR2.2; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA DP14-beta chain; HLA-D; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DR; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; lymphocyte antigen DRB1; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DR beta 1; MHC Class 2; MHC class II; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB | |
Mouse | |
Protein A | |
RUO | |
3115, 3123 | |
4° C, do not freeze | |
Liquid |