Learn More
MIF Recombinant Polyclonal Antibody (7HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 710240
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with non-human primate, mouse and rat based on sequence homology. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
MIF Recombinant Polyclonal antibody specifically detects MIF in Human samples. It is validated for Western Blot.Specifications
MIF | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
cytokine; delayed early response protein 6; DER6; GIF; GLIF; glutathione-binding 13 kDa protein; Glycosylation-inhibiting factor; HGNC:7097; L-dopachrome isomerase; L-dopachrome tautomerase; macrophage migration inhibitory factor; macrophage migration inhibitory factor (glycosylation-inhibiting factor); MGC107654; Mif; MMIF; phenylpyruvate tautomerase | |
Recombinant protein corresponding to amino acids 2–115 of human MIF | |
100 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
7HCLC | |
Unconjugated | |
P14174 | |
Rabbit | |
Protein A | |
RUO | |
4282 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |