missing translation for 'onlineSavingsMsg'
Learn More

MIP-3 alpha, PE, clone: 67310, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Thermo Scientific Pierce MA523583

Additional Details : Weight : 0.00010kg

Product Code. 15563272

  • 2860.00 SEK / 100 tests


Estimated Shipment date
from Supplier 08-05-2019
Add to basket

Description

Description

In ELISAs, this antibody does not cross-react with recombinant human (rh) CCL19, rhCXCL1, rhCCL16, rhCCL11, recombinant mouse CCL20, or recombinant rat CCL20.
Specifications

Specifications

MIP-3 alpha/CCL20
Monoclonal
PE
CCL20
Purified
CCL20
E. coli-derived recombinant human CCL20/MIP-3 alpha Ala27-Met96 (Asn95Asp)
Protein A/G
Store at 4°C, store in the dark, do not freeze
Primary
6364
Flow Cytometry
67310
Buffered saline solution with BSA and 0.09% sodium azide
P78556
CKb4, LARC, MIP-3a, MIP3A, SCYA20, ST38, C-C motif chemokine 20, CC chemokine LARC, MIP-3-alpha, beta chemokine exodus-1, beta-chemokine exodus-1, exodus-1, liver and activation-regulated chemokine, macrophage inflammatory protein 3 alpha, small inducible cytokine subfamily A (Cys-Cys), member 20, small-inducible cytokine A20
Murine
IgG1
100 Tests
Antibody
Monoclonal
Human
Videos