missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIS12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544882
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TLTFFDELHN VGRDHGTSDF RESLVSLVQN SRKLQNIRDN VEKESKRLKI Sequence homology: Cow: 86%; Horse: 86%; Human: 100%; Mouse: 91%; Rabbit: 86%; Rat: 100%
MIS12 Polyclonal antibody specifically detects MIS12 in Human samples. It is validated for Western Blot.Specifications
MIS12 | |
Polyclonal | |
Unconjugated | |
MIS12 | |
2510025F08Rik; Ac1585; AU044651; hMis12; homolog of yeast Mis12; KNTC2AP; Liver regeneration-related protein LRRG112; Mis12; MIS12 homolog; MIS12 homolog (yeast); MIS12 kinetochore complex component; MIS12, kinetochore complex component; MIS12, MIND kinetochore complex component, homolog; Misc12; MTW1; Protein MIS12 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79003 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H081 | |
MIS12 | |
Synthetic peptide directed towards the middle region of human MIS12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |