Learn More
SLC25A33 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521793
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (87%), Rat (88%), Sheep (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SLC25A33 Polyclonal antibody specifically detects SLC25A33 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SLC25A33 | |
Polyclonal | |
Unconjugated | |
SLC25A33 | |
BMSC-MCP; Bone marrow stromal cell mitochondrial carrier protein; huBMSC-MCP; mitochondrial carrier protein; novel mitochondrial carrier protein; PNC1; PNC1 protein; Protein PNC1; SLC25A33; solute carrier family 25 (pyrimidine nucleotide carrier), member 33; solute carrier family 25 member 33; solute carrier family 25, member 33 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84275 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9BSK2 | |
SLC25A33 | |
Recombinant fragment corresponding to a region within amino acids 114 and 321 of SLC25A33 (Uniprot ID#Q9BSK2) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |