Learn More
Goat anti-Mouse IgE Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184764
2763.47 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Goat anti mouse IgE polyclonal antibody recognizes mouse IgE and shows negligible cross - reactivity with other immunoglobulins.
Mouse IgE Polyclonal antibody specifically detects Mouse IgE in Mouse samples. It is validated for ELISA.Specifications
Mouse IgE | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Native mouse IgE. | |
500 μg | |
Secondary | |
4° C, do not freeze | |
IgG |