missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Mouse IgG peroxidase-linked whole antibody (from sheep) Secondary Antibody, GE Healthcare
Brand: Life Science products Cytiva 25005363
This item is not returnable.
View return policy
Description
Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms
Protein Blotting, ELISA, Immunocytochemistry
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?