Learn More
MPHOSPH6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530364
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Human (99%), Mouse (88%), Rat (88%), Chicken (81%), Rhesus Monkey (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MPHOSPH6 Polyclonal antibody specifically detects MPHOSPH6 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MPHOSPH6 | |
Polyclonal | |
Unconjugated | |
MPHOSPH6 | |
1110001M01Rik; AA536809; C86426; M phase phosphoprotein 6; M-phase phosphoprotein 6; Mphosph6; MPP; MPP6; MPP-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10200 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.77 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q99547 | |
MPHOSPH6 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 130 of Human MPHOSPH6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |