missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRE11 Mouse anti-Human, Clone: 44B12, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517261
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is HeLa cell lysate.
MRE11 (double-strand break repair protein MRE11) is a component of the MRN complex. This complex plays a central role in double-strand break repair, DNA recombination, maintece of telomere integrity and meiosis. The complex possesses single-stranded endonuclease activity and double-strand-specific 3′-5′ exonuclease activity which are provided by MRE11. RAD50 is required to bind DNA ends and hold them in close proximity. Mutations in the gene could result in ataxia-telangiectasia-like disorder 1.Specifications
MRE11 | |
Monoclonal | |
1 mg/mL | |
HEPES with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide | |
P49959 | |
MRE11 | |
Recombit human protein purified from E.coli (His-MRE11) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunoprecipitation, Western Blot | |
44B12 | |
Unconjugated | |
MRE11 | |
AT-like disease; DNA recombination and repair protein; MRE11 homolog 1; MRE11 homolog A; double-strand break repair protein MRE11A; endo/exonuclease Mre11; meiotic recombination 11 homolog 1; meiotic recombination 11 homolog A | |
Mouse | |
Protein G | |
RUO | |
4361 | |
-20°C | |
Liquid |