missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRE11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531262
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, A431, C8D30, NIH-3T3, PC-12. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MRE11 Polyclonal antibody specifically detects MRE11 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
MRE11 | |
Polyclonal | |
Unconjugated | |
Mre11 | |
ATLD; AT-like disease; DNA recombination and repair protein; double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; HNGS1; Meiotic recombination 11 homolog 1; meiotic recombination 11 homolog A; meiotic recombination 11 homolog A (S. cerevisiae); mmMRE11A; MRE11; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog A, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11 homolog A (S. cerevisiae); MRE11 meiotic recombination 11-like protein A; MRE11A; MRE11B; MRX complex nuclease subunit; NGS1; RAD58; XRS4; YM9959.06C; YMR224C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17535, 4361, 64046 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.57 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P49959, Q61216, Q9JIM0 | |
Mre11, Mre11a | |
Recombinant protein fragment corresponding to a region within amino acids 132 and 572 of Human Mre11 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |