Learn More
MRPS22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528478
Additional Details : Weight : 0.01000kg
Description
PA5-28478 targets MRPS22 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28478 immunogen is recombit fragment corresponding to a region within amino acids 165 and 360 of Human MRPS22. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that does not seem to have a counterpart in prokaryotic and fungal-mitochondrial ribosomes. This gene lies telomeric of and is transcribed in the opposite direction from the forkhead box L2 gene. A pseudogene corresponding to this gene is found on chromosome Xq.Specifications
MRPS22 | |
Polyclonal | |
Unconjugated | |
P82650 | |
MRPS22 | |
Recombit fragment corresponding to a region within amino acids 165 and 360 of Human MRPS22 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
MRPS22 | |
C3orf5, COXPD5, GIBT, MRP-S22, RPMS22, 28S ribosomal protein S22, mitochondrial, S22mt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56945 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |