Learn More
mtHSP70 Monoclonal Antibody (JG1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA3028
Additional Details : Weight : 1.24740kg
Description
MA3-028 detects mitochondrial heat shock protein 70 kDa (mtHSP70) from human, non-human primate, canine and mouse tissues. MA3-028 has been successfully used in Western blot, immunocytochemical, immunofluorescence, immunohistochemical (paraffin), and immunoprecipitation procedures. By Western blot, this antibody detects a single ∽75 kDa band representing mtHSP70 from U2OS cell homogenate. Immunocytochemical staining of mtHSP70 in DAP.3 cells with MA3-028 results in a worm-like staining pattern, consistent with mitochondrial localization. Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay.
mtHSP70 Monoclonal antibody specifically detects mtHSP70 in Canine, Human, Mouse, Non-human Primate samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
mtHSP70 | |
Monoclonal | |
1.07 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P0DMV8, Q61696 | |
HSPA1A | |
Synthetic peptide corresponding to residues G(615) S G S S G T G E Q K E D Q K E E K Q(633) of mouse mtHSP70. | |
100 μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate | |
Antibody | |
IgG3 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
JG1 | |
Unconjugated | |
HSPA1A | |
68 kDa heat shock protein; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 1A; heat shock 70kDa protein 1A; heat shock protein 1A; heat shock protein family A (Hsp70) member 1A; heat shock protein, 70 kDa 3; heat shock-induced protein; HEL-S-103; Hsp68; HSP70.1; HSP70.1/HSP70.2; Hsp70.3; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; HSP70-2; Hsp70-3; Hsp70A1; HSP70I; Hsp72; HSPA1; Hspa1a; HSPA1B; HSX70; inducible heat shock protein 70 | |
Mouse | |
Protein A | |
RUO | |
193740, 3303 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |