missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513711
Additional Details : Weight : 0.01000kg
Description
MTMR4 Polyclonal antibody specifically detects MTMR4 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
MTMR4 | |
Polyclonal | |
Unconjugated | |
MTMR4 | |
AA596759; DKFZp781E194; ESTM44; FYVE domain-containing dual specificity protein phosphatase 2; FYVE zinc finger phosphatase; FYVE-DSP2; KIAA0647; MGC163449; MGC163451; mKIAA0647; Mtmr4; MTMR7; myotubularin related protein 4; myotubularin-related protein 4; ZFYVE11; Zinc finger FYVE domain-containing protein 11; zinc finger, FYVE domain containing 11 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
170749, 9110 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q91XS1, Q9NYA4 | |
MTMR4 | |
KLH conjugated synthetic peptide between 1155-1187 amino acids from the C-terminal region of human MTMR4 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |