Learn More
CRYM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528198
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Mouse brain. Predicted reactivity: Mouse (90%), Rat (90%), Rhesus Monkey (99%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CRYM Polyclonal antibody specifically detects CRYM in Human, Mouse samples. It is validated for Western Blot.Specifications
CRYM | |
Polyclonal | |
Unconjugated | |
CRYM | |
CDK108; Crym; crystallin mu; crystallin, mu; DFNA40; ketimine reductase mu-crystallin; mu-crystallin homolog; Mu-crystallin-like protein; NADP-regulated thyroid-hormone binding protein; NADP-regulated thyroid-hormone-binding protein; similar to Mu-crystallin homolog (NADP-regulated thyroid-hormone binding protein); THBP; thiomorpholine-carboxylate dehydrogenase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12971, 1428 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.7 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O54983, Q14894 | |
CRYM | |
Recombinant fragment corresponding to a region within amino acids 43 and 314 of Human mu Crystallin | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |