missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPML1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen OSM00035W
Additional Details : Weight : 0.01000kg
Description
OSM00035W is specific for Mucolipin 1, isoform 1.
Specifications
TRPML1 | |
Polyclonal | |
MCOLN1 | |
Mucolipin-1, Mucolipidin, Mcoln1, Mucolipin 1, Mcoln1, MCLN1, TRPML1, MGC7172, mucolipidin, 2210015I05Rik, Mucolipin-1, MG-2, ML4, MSTP080, MLIV, TRP-ML1, TRPM-L1, MCOLN1 mucolipin 1 | |
Sheep | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9GZU1 | |
MCOLN1 | |
A synthetic peptide from human Mucolipin 1 | |
RUO | |
57192 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |