Learn More
MUNC18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1742
Additional Details : Weight : 0.01000kg
Description
PA1-742 detects Munc-18 from human, rat and mouse tissues. PA1-742 has been successfully used in Western blot, immunohistochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects an ∽67 kDa protein representing Munc-18 from rat and mouse whole brain extract. PA1-742 immunizing peptide corresponds to amino acid residues 58-70 from human Munc-18. This sequence is completely conserved between human, mouse, rat, bovine, and canine Munc-18. PA1-742 immunizing peptide (Cat. # PEP-067) is available for use in neutralization and control experiments.
MUNC18 Polyclonal antibody specifically detects MUNC18 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
MUNC18 | |
Polyclonal | |
Unconjugated | |
Stxbp1 | |
AI317162; AI326233; ANC18HA; EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; MMS10-G; Ms10g; MUNC18-1; Munc-18a; munc-18c; neuronal SEC1; NSEC1; n-sec1; NSEC1A; p67; Protein unc-18 homolog 1; protein unc-18 homolog A; rbSec1; Rb-sec1; rbSec1A; rbSec1B; rop; RP11-56D16.3; Sec1; Stxbp1; Sxtbp1; syntaxin binding protein 1; syntaxin-binding protein 1; UNC18; UNC-18 homolog; unc-18-1; Unc18-1; UNC18A; unc-18A; Unc18h | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20910, 25558, 6812 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
O08599, P61764, P61765 | |
Stxbp1 | |
Synthetic Peptide: V(58) E D I N K R R E P I P S(70) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |