Learn More
MSI2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531024
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: IMR32, MCF-7, mouse brain. Predicted reactivity: Mouse (100%), Xenopus laevis (97%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MSI2 Polyclonal antibody specifically detects MSI2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
MSI2 | |
Polyclonal | |
Unconjugated | |
MSI2 | |
1700105C15Rik; AI451722; AI563628; AW489193; MSI2; Msi2h; musashi homolog 2; musashi RNA binding protein 2; musashi RNA-binding protein 2; Musashi2; musashi-2; RGD1560397; RNA-binding protein Musashi homolog 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124540, 360596, 76626 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q920Q6, Q96DH6 | |
MSI2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 217 of Human Musashi 2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |