missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MYO1C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532232
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: A431, MYO1C transfected 293T lysate. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (80%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MYO1C is a member of the unconventional myosin protein family, which are actin-based molecular motors. The protein is found in the cytoplasm, and one isoform with a unique N-terminus is also found in the nucleus. The nuclear isoform associates with RNA polymerase I and II and functions in transcription initiation. The mouse ortholog of this protein also functions in intracellular vesicle transport to the plasma membrane.
Specifikationer
| MYO1C | |
| Polyclonal | |
| Unconjugated | |
| MYO1C | |
| C80397; FLJ23903; mm1beta; MMIb; MMI-beta; myo1c; myo1ca; MYO1E; myosin 1c, paralog a; myosin heavy chain myr 2; myosin I; Myosin I beta; myosin IB; myosin IC; myosin Ic, paralog a; myosin IC-like; myosin-I beta; myosin-Ibeta; myosin-Ic; Myr2; NMI; nuclear myosin I; nuclear myosin I beta; si:ch211-69i14.4; unconventional myosin Myr2 I heavy chain; unconventional myosin-Ic | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4641 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O00159 | |
| MYO1C | |
| Recombinant protein fragment corresponding to a region within amino acids 1 and 1028 of Human Myosin 1C | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering