missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ NCF4 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 13493117
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
13493117 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 13493117 Leverantör Invitrogen™ Leverantörsnummer PA529000

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji, K562, THP-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a cytosolic regulatory component of the superoxide-producing phagocyte NADPH-oxidase, a multicomponent enzyme system important for host defense. This protein is preferentially expressed in cells of myeloid lineage. It interacts primarily with neutrophil cytosolic factor 2 (NCF2/p67-phox) to form a complex with neutrophil cytosolic factor 1 (NCF1/p47-phox), which further interacts with the small G protein RAC1 and translocates to the membrane upon cell stimulation. This complex then activates flavocytochrome b, the membrane-integrated catalytic core of the enzyme system. The PX domain of this protein can bind phospholipid products of the PI(3) kinase, which suggests its role in PI(3) kinase-mediated signaling events. The phosphorylation of this protein was found to negatively regulate the enzyme activity. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen NCF4
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 0.87 mg/mL
Konjugera Unconjugated
Formulering 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gen Ncf4
Genaccessionsnr. Q15080
Gene Alias AI451400; CGD3; CTA-833B7.1; MGC3810; NCF; Ncf4; NCF-4; Neutrophil cytosol factor 4; neutrophil cytosolic factor 4; neutrophil cytosolic factor 4, 40kDa; neutrophil NADPH oxidase factor 4; OTTHUMP00000028736; OTTHUMP00000043756; P40PHOX; p40-phox; SH3 and PX domain-containing protein 4; SH3PXD4
Gensymboler Ncf4
Värd art Rabbit
Immunogen Full length human NCF4 Recombinant protein.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 4689
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.