missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEFM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA316720
Additional Details : Weight : 0.01000kg
Description
This antibody is likely to react with most mammals. Suggested positive control: antigen standard for NEFM (transient overexpression lysate).
NEFM Polyclonal antibody specifically detects NEFM in Bovine, Chicken, Equine, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
NEFM | |
Polyclonal | |
Unconjugated | |
NEFM | |
160 kDa neurofilament protein; major neuronal intermediate filament subunit; medium neurofilament subunit; middle molecular weight neurofilament protein NF-M(2); Nef3; Nefm; nef-m; nefm.S; nefm-a; nefm-b; Neurofilament 3; neurofilament 3 (150kDa medium); neurofilament 3, medium; neurofilament M subunit; neurofilament medium; neurofilament medium polypeptide; neurofilament medium S homeolog; neurofilament medium tail domain; neurofilament protein M; Neurofilament protein, middle polypeptide; neurofilament triplet M protein; neurofilament, medium polypeptide; neurofilament, medium polypeptide 150kDa; neurofilament, medium polypeptide S homeolog; neurofilament-3 (150 kD medium); neurofilament-M; neurofilament-M subunit; neuronal intermediate filament; NF160; NF165; NFM; NF-M; NF-M c-terminus; NF-M protein; NMC; XELAEV_18020229mg | |
Rabbit | |
100 μL | |
Primary | |
Bovine, Chicken, Equine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 5mM sodium azide | |
O77788, P07197, P08552, P08553, P12839, P16053 | |
NEFM | |
Recombinant fusion protein containing the extreme C-terminal segment of rat NF-M, amino acids 549-845 | |
RUO | |
100054129, 100625407, 18040, 24588, 281347, 396206, 4741 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |