missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ NFS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540653
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide sequence: TTQTEHKCVL DSCRSLEAEG FQVTYLPVQK SGIIDLKELE AAIQPDTSLV Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Yeast: 92%.
NFS1 (nitrogen fixation 1), also known as NIFS or IscS (cysteine desulfurase), is a member of the class V pyridoxal-phosphate-dependent aminotransferase family. It localizes to the cytoplasm or mitochondrion depending on which form is generated based on cytosolic pH. Highest expression levels of NFS1 are found in heart and skeletal muscle. Lower levels of expression are also found in liver, brain and pancreas. NFS1 is responsible for catalyzing the removal of sulfur from cysteine to form alanine, thereby supplying the inorganic sulfur for iron-sulfur (Fe-S) clusters. Fe-S clusters function as essential cofactors in a wide variety of events, including facilitation of electron transfer processes in oxidative phosphorylation, catalysis of enzymatic reactions in aconitase and dehydratases and maintenance of structural integrity in the DNA repair enzyme endonuclease III.
Specifikationer
| NFS1 | |
| Polyclonal | |
| Unconjugated | |
| NFS1 | |
| AA987187; cysteine desulfurase, mitochondrial; Cysteine desulfurase, mitochondrial-like protein; HUSSY-08; IscS; LOW QUALITY PROTEIN: cysteine desulfurase, mitochondrial; m-Nfs1; m-Nfsl; Nfs1; NFS1 cysteine desulfurase; NFS1 nitrogen fixation 1 homolog; NFS1 nitrogen fixation 1 homolog (S. cerevisiae); NFS1, cysteine desulfurase; Nifs; nifS-like (sic); nitrogen fixation 1 (S. cerevisiae, homolog); nitrogen fixation gene 1; nitrogen fixation gene 1 (S. cerevisiae); nitrogen fixation gene, yeast homolog 1; nitrogen-fixing bacteria S-like protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 9054 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y697 | |
| NFS1 | |
| Synthetic peptide directed towards the middle region of human NFS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering