Learn More
NKRF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528242
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (97%), Chicken (84%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NKRF Polyclonal antibody specifically detects NKRF in Human samples. It is validated for Western Blot.Specifications
NKRF | |
Polyclonal | |
Unconjugated | |
NKRF | |
9430034D17Rik; AW124770; C80287; ITBA4; ITBA4 protein; LOC100910054; NF-kappaB repressing factor; NF-kappa-B-repressing factor; NF-kappa-B-repressing factor-like; NFKB repressing factor; NFkB-repressing factor; NKRF; NRF; Protein ITBA4; Transcription factor NRF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55922 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O15226 | |
NKRF | |
Recombinant fragment corresponding to a region within amino acids 335 and 690 of Human NKRF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |