missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518551
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine, human and mouse based on sequence homology.
NMDAR1 Polyclonal antibody specifically detects NMDAR1 in Human, Mouse samples. It is validated for Western Blot.Specifications
NMDAR1 | |
Polyclonal | |
Unconjugated | |
GRIN1 | |
GluN1; Glun1r; GluRdelta1; Glurz1; GluRzeta1; glutamate [NMDA] receptor subunit zeta 1; glutamate [NMDA] receptor subunit zeta-1; glutamate ionotropic receptor NMDA type subunit 1; glutamate receptor; glutamate receptor ionotropic, NMDA 1; glutamate receptor subunit zeta 1; glutamate receptor, ionotropic, NMDA1 (zeta 1); glutamate receptor, ionotropic, N-methyl D-aspartate 1; GRIN1; M100174; MRD8; neurotransmitter receptor; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDA1; Nmdar; NMDAR1; NMD-R1; NMDZ1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMZ1; NR1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; Rgsc174; RP11-350O14.1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14810, 2902 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P35438, Q05586 | |
GRIN1 | |
Synthetic peptide sequence (TQERVNNSNKKE) corresponding to the internal amino acids of GRIN1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |