missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533143
Additional Details : Weight : 0.01000kg
Description
Rabbit anti Human non-neuronal enolase polyclonal antibody recognizes human non-neuronal enolase, also known as alpha enolase or enolase 1.
ENO1 Polyclonal antibody specifically detects ENO1 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
ENO1 | |
Polyclonal | |
Unconjugated | |
ENO1 | |
0610008I15; 2-phospho-D-glycerate hydrolase; 2-phospho-D-glycerate hydro-lyase; AL022784; alpha enolase; alpha enolase like 1; alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; c-Myc promoter binding protein; C-myc promoter-binding protein; Eno1; Eno-1; ENO1L1; enoa; Enolase 1; enolase 1, (alpha); enolase 1, alpha non-neuron; enolase-alpha; epididymis secretory protein Li 17; HEL-S-17; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin | |
Rabbit | |
Protein G | |
RUO | |
2023 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
5 mg/mL | |
PBS with 0.09% sodium azide | |
P06733 | |
ENO1 | |
Native human NNE | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |