missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOS1AP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523774
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
NOS1AP Polyclonal antibody specifically detects NOS1AP in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
NOS1AP | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O75052 | |
NOS1AP | |
KLH conjugated synthetic peptide between 437-466 amino acids from the central region of human NOS1AP | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
NOS1AP | |
6330408P19Rik; Capon; carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein; C-terminal PDZ domain ligand of neuronal nitric oxide synthase; C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON); C-terminal PDZ ligand of neuronal nitric oxide synthase protein; Kiaa0464; ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain; MGC138500; nitric oxide synthase 1 (neuronal) adaptor protein; nitric oxide synthase 1 adaptor protein; NOS1AP | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
9722 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |