missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTCH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523455
Additional Details : Weight : 0.01000kg
Description
NOTCH2 Polyclonal antibody specifically detects NOTCH2 in Human samples. It is validated for Western Blot.Specifications
NOTCH2 | |
Polyclonal | |
Unconjugated | |
NOTCH2 | |
AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2 | |
Rabbit | |
Protein A | |
RUO | |
4853 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q04721 | |
NOTCH2 | |
A portion of amino acids 1550-1600 of human NOTCH2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |