missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOX2 Monoclonal Antibody (NL7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518051
Additional Details : Weight : 0.01000kg
Description
This antibody is specific for NOX2/gp91phox (aa 498-507, EKDVITGRKQ).
Mox1 and the glycoprotein gp91-phox are largely related proteins that are essential components of the NADPH oxidase. The superoxide-generating NADPH oxidase complex expresses in phagocytes, neuroepithelial bodies, vascular smooth muscle cells, and endothelial cells. It includes a membrane bound flavocytochrome containing two subunits, gp91-phox and p22-phox, and the cytosolic proteins p47-phox and p67-phox. The p22- and gp91-phox subunits also function as surface O2 sensors that initiate cellular signaling in response to hypoxic conditions. Mox1 and gp91 contain identical C-terminal sequence identity, yet possess distinct expression patterns. gp91-phox expresses in eosinophils, neutrophils, monocytes, and B-lymphocytes, whereas Mox1 is predominantly detected in the colon, with low expression in the uterus and prostate.Specifications
NOX2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P04839 | |
Cybb | |
Partially purified human neutrophil flavocytochrome b (heparin Ultrogellectin affinity purification in octyl glucoside) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
NL7 | |
Unconjugated | |
Cybb | |
AMCBX2; C88302; Cgd; CGD91-phox; Cybb; Cyd; cytochrome b(558) subunit beta; cytochrome b-245 beta chain; cytochrome b-245 beta polypeptide; cytochrome b-245 heavy chain; cytochrome b-245, beta polypeptide; cytochrome b558 subunit beta; endothelial type gp91-phox; GP91 PHOX; GP911; GP91-1; GP91PHOX; gp91-phox; Heme-binding membrane glycoprotein gp91phox; IMD34; NADPH oxidase 2; neutrophil cytochrome b 91 kDa polypeptide; Nox2; p22 phagocyte B-cytochrome; p91PHOX; p91-PHOX; superoxide-generating NADPH oxidase heavy chain subunit | |
Mouse | |
Protein G | |
RUO | |
1536 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |