missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOX5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543433
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: DQAEEAQYGR FLELHMYMTS ALGKNDMKAI GLQMALDLLA NKEKKDSITG Sequence homology: Cow: 86%; Human: 100%; Pig: 79%; Rabbit: 100%
NOX5 Polyclonal antibody specifically detects NOX5 in Human samples. It is validated for Western Blot.Specifications
NOX5 | |
Polyclonal | |
Unconjugated | |
NOX5 | |
NADPH oxidase 5; NADPH oxidase, EF-hand calcium binding domain 5; NOX5; NOX5zeta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79400 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96PH1 | |
NOX5 | |
Synthetic peptide directed towards the C-terminal of human NOX5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |