missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfactory Marker Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSR00038W
Additional Details : Weight : 0.01000kg
Description
In IHC applications, Zamboni's or 4% PFA fixatives are recommended.
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.Specifications
Olfactory Marker Protein | |
Polyclonal | |
Olfactory neuronal-specific protein; OMP | |
Synthetic peptide from hamster Olfactory Marker Protein. | |
RUO | |
Hamster | |
Antibody | |
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Rabbit | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |