missing translation for 'onlineSavingsMsg'
Learn More

OLLAS Tag Monoclonal Antibody (L2), Invitrogen™

Rat Monoclonal Antibody

Brand:  Invitrogen MA516125

Additional Details : Weight : 0.10000kg

Product Code. 13217598

  • 4430.00 SEK / 500µL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

OLLAS Tag Monoclonal antibody specifically detects OLLAS Tag in Tag samples. It is validated for ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications

Specifications

OLLAS Tag
Monoclonal
1.0 mg/mL
PBS with 0.05% sodium azide
OLLAS (SGFANELGPRLMGK)-tagged extracellular domain of mouse Langerin
100 μL
Primary
Antibody
IgG1 κ
ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
L2
Unconjugated
Rat
Protein G
RUO
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers