missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Oxytocin Receptor Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534065
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Mouse, Elephant, Panda, Pig (95%).
The oxytocin receptor (OXTR) belongs to the G-protein coupled receptor family. Its activity is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. The receptor is made up of 389 amino acids and has a molecular weight of ~43,000 kDa. The oxytocin-oxytocin receptor system plays an important role as an inducer of uterine contractions during parturition and of milk ejection. Oxytocin receptors are expressed by the myoepithelial cells of the mammary gland, and in both the myometrium and endometrium or the uterus at the end of pregnancy. Additionally, oxytocin receptors are present in the central nervous system, mediating a variety of behaviors, including maternal behavior, bonding, social memory and recognition, sexual and aggressive behaviors, and stress and anxiety.
Specifikationer
| Oxytocin Receptor | |
| Polyclonal | |
| Unconjugated | |
| Oxtr | |
| Otr; OT-R; OTR1; oxcytocin receptor; OXTR; Oxt-r; Oxytocin receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5021 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P30559 | |
| Oxtr | |
| Synthetic 20 amino acid peptide from N-terminal extracellular domain of human Oxytocin Receptor. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering