missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p400 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530780
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: MCF-7. Predicted reactivity: Mouse (83%), Rat (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
p400 Polyclonal antibody specifically detects p400 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
p400 | |
Polyclonal | |
Unconjugated | |
EP400 | |
1700020J09Rik; AU023439; CAG repeat protein 32; CAGH32; Domino homolog; E1A binding protein p400; E1A-binding protein p400; EP400; hDomino; KIAA1498; KIAA1818; mDomino; mKIAA1498; P400; p400 kDa SWI2/SNF2-related protein; p400 SWI2/SNF2-related protein; TNRC12; trinucleotide repeat containing 12; trinucleotide repeat-containing gene 12 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57634 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.95 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96L91 | |
EP400 | |
Recombinant fragment corresponding to a region within amino acids 136 and 430 of Human p400 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |