missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRB Monoclonal Antibody (K.596.7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515103
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with PDGF receptor -alpha.
PDGFRB Monoclonal antibody specifically detects PDGFRB in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western Blot.Specifications
PDGFRB | |
Monoclonal | |
1.75 mg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P05622, P09619, Q05030 | |
Pdgfrb | |
Recombinant human PDGF receptor β protein | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
K.596.7 | |
Unconjugated | |
Pdgfrb | |
AI528809; Beta platelet-derived growth factor receptor; beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140B; CD140b antigen; IBGC4; IMF1; JTK12; KOGS; PDGF beta chain; PDGF Receptor beta; Pdgfr; Pdgfr1; PDGFR-1; PDGFRB; PDGF-R-beta; PDGFR-beta; PENTT; platelet derived growth factor receptor beta; platelet derived growth factor receptor, beta polypeptide; platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide | |
Mouse | |
Affinity chromatography | |
RUO | |
18596, 24629, 5159 | |
-20°C | |
Liquid |