missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Phospho-4EBP1 (Thr37) Recombinant Rabbit Monoclonal Antibody (52H37L2)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ 700238

Produktkod. 15326954

  • 4505.00 SEK / 100µg

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!


For Research Use Only. All usage must comply with product instructions.

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

This antibody is predicted to react with bovine, equine, mouse, opossum, rat and zebrafish based on sequence homology. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

Phospho-4EBP1 (Thr37)
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
Q13541
EIF4EBP1
A peptide corresponding to amino acids 31-41 of Q13541.
100 μg
Primary
Human
Antibody
IgG
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
52H37L2
Unconjugated
EIF4EBP1
4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1
Rabbit
Protein A
RUO
1978
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.