missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-alpha Adducin (Ser726) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538064
Additional Details : Weight : 0.00900kg
Description
Adducins are a family of cytoskeletal proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha and beta adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha and gamma adducins are ubiquitously expressed. In contrast, beta adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca2+/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.Specifications
Phospho-alpha Adducin (Ser726) | |
Polyclonal | |
Unconjugated | |
ADD1 | |
ADDA, Alpha adducin, Erythrocyte adducin alpha subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11518, 118, 24170 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35611, Q9QYC0, Q63028 | |
ADD1 | |
A synthetic phosphopeptide derived from human ADD1around the phosphorylation site of Ser726 (T-P-SP-F-L) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |