Learn More
Phospho-CREB (Ser133) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14619
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, chicken, human, mouse, non-human primate, rat, sheep, Xenopus and zebrafish based on 100% sequence homology. This antibody is specific for the ∽45 kDa CREB phosphorylated at Ser133 in Western blots of rat brain lysates. Immunolabeling can be blocked by lambda-phosphatase treatment.
Phospho-CREB (Ser133) Polyclonal antibody specifically detects Phospho-CREB (Ser133) in Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Phospho-CREB (Ser133) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P15337, Q01147 | |
CREB1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser133 of rat CREB1 conjugated to KLH | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
CREB1 | |
2310001E10Rik; 3526402H21Rik; active transcription factor CREB; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CREB; CREB1; Creb-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; htCREB; MGC9284; OTTHUMP00000206661; OTTHUMP00000206662; OTTHUMP00000206667; transactivator protein; Y protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
12912, 81646 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |