missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-EGFR (Tyr1086) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517281
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other EGFR family members.
Phospho-EGFR (Tyr1086) Polyclonal antibody specifically detects Phospho-EGFR (Tyr1086) in Canine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Phospho-EGFR (Tyr1086) | |
Polyclonal | |
Unconjugated | |
EGFR | |
2.7.10.1; 9030024J15Rik; AI552599; avian erythroblastic leukemia viral (v-erbB) oncogene homolog; avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; EC 2.7.10.1; egf receptor; Egfr; EGFR-related peptide; epidermal growth factor receptor; epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian); Epidermal growth factor receptor formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); epidermal growth factor receptor, formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); ERBB; ERBB1; ErbB-1; erb-b2 receptor tyrosine kinase 1; Errb1; Errp; HER1; kinase EGFR; mENA; NISBD2; Oncogene ERBB; PIG61; Proto-oncogene c-ErbB-1; receptor tyrosine-protein kinase erbB-1; Urogastrone; wa2; wa-2; Wa5; waved 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13649, 1956, 24329, 404306 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
81 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P00533, Q01279 | |
EGFR | |
Synthetic phosphopeptide corresponding to residues surrounding pTyr1086 of human EGF receptor | |
100 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG |