missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-G3BP1 (Ser232) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537781
Additional Details : Weight : 0.00910kg
Description
A suggested positive control for Western blot is 293 cells; suggested positive control for IHC is human breast carcinoma; suggested positive control for ICC/IF is Hela cells.
Phospho-G3BP1 (Ser232) Polyclonal antibody specifically detects Phospho-G3BP1 (Ser232) in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-G3BP1 (Ser232) | |
Polyclonal | |
Unconjugated | |
G3BP1 | |
AI849976; ATP-dependent DNA helicase VIII; B430204O07; C87777; DNA helicase VIII; G3BP; G3BP stress granule assembly factor 1; G3bp1; G3BP-1; GAP binding protein; GAP SH3 binding protein; GAP SH3 domain-binding protein 1; GTPase activating protein (SH3 domain) binding protein 1; hDH VIII; HDH-VIII; MGC111040; mKIAA4115; Ras GTPase-activating protein-binding protein 1; RasGAP-associated endoribonuclease G3BP; Ras-GTPase-activating protein SH3-domain binding protein 1; Ras-GTPase-activating protein SH3-domain-binding protein; RGD1310666 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10146 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13283 | |
G3BP1 | |
Peptide sequence around phosphorylation site of serine 232 (S-S-S(p)-P-A) derived from Human G3BP-1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |