missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IRS1 (Ser794) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536752
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 132 and 180 kDa. Purity is >95% by SDS-PAGE.
Phospho-IRS1 (Ser794) Polyclonal antibody specifically detects Phospho-IRS1 (Ser794) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Phospho-IRS1 (Ser794) | |
Polyclonal | |
Unconjugated | |
IRS1 | |
G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16367, 25467, 3667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P35568, P35569, P35570 | |
IRS1 | |
Synthetic phosphopeptide derived from human IRS-1 around the phosphorylation site of Serine 794 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |