missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PDGFRA (Tyr742) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 441006
Additional Details : Weight : 0.02000kg
Description
Phospho-PDGFRA (Tyr742) Polyclonal antibody specifically detects Phospho-PDGFRA (Tyr742) in Human, Mouse samples. It is validated for Western Blot.Specifications
Phospho-PDGFRA (Tyr742) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P16234, P26618 | |
PDGFRA | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human PDGFR-alpha that contains tyrosine 742. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PDGFRA | |
AI115593; alpha platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); alpha-type platelet-derived growth factor receptor; APDGFR; CD140 antigen-like family member A; CD140A; CD140a antigen; GAS9; MGC74795; OTTHUMP00000218656; PDGF A-chain; PDGF alpha chain; PDGF Receptor alpha; PDGF subunit A; PDGF-1; PDGFACE; PDGFR2; Pdgfr-2; PDGFRA; pdgfr-a; PDGFRA/BCR fusion; PDGF-R-alpha; PDGFR-alpha; platelet derived growth factor receptor alpha; platelet derived growth factor receptor, alpha polypeptide; platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 2; platelet-derived growth factor receptor alpha; platelet-derived growth factor receptor, alpha polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein; RHEPDGFRA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18595, 5156 | |
-20°C | |
Liquid |