missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-SMAD1 (Ser187) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536876
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 60 kDa. Purity is >95% by SDS-PAGE.
Phospho-SMAD1 (Ser187) Polyclonal antibody specifically detects Phospho-SMAD1 (Ser187) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Phospho-SMAD1 (Ser187) | |
Polyclonal | |
Unconjugated | |
SMAD1 | |
AI528653; BSP1; BSP-1; dwarfin-A; Dwf-A; DWFC; hSMAD1; JV41; JV4-1; JV5-1; MAD (mothers against decapentaplegic, Drosophila) homolog 1; MAD homolog 1; MAD homolog1 (mothers against decapentaplegic, Drosophila); MAD, mothers against decapentaplegic homolog 1; Madh1; MADH5; MADR1; Mad-related protein 1; Mlp1; mMad1; mothers against decapentaplegic homolog 1; mothers against DPP homolog 1; mothers-against-DPP-related 1; Mothers-against-DPP-related-1; MusMLP; OTTHUMP00000220456; SMAD; Smad 1; SMAD family member 1; SMAD, mothers against DPP homolog 1; Smad1; SMAD5; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17125, 25671, 4086 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P70340, P97588, Q15797 | |
SMAD1 | |
Synthetic phosphopeptide derived from human Smad1 around the phosphorylation site of Serine 187 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |